2,2'-Bithiophene 99%

Supplier: Apollo Scientific
Warning

Synonyms: 2,2′-Dithienyl, 2,2′-Bithienyl

OR1311-100G OR1311-25G OR1311-500G
APOSOR1311-100GEA 60.9 EUR
APOSOR1311-100G APOSOR1311-25G APOSOR1311-500G
2,2'-Bithiophene 99%
2,2'-Bithiophene
Formula: C₈H₆S₂
MW: 166.27 g/mol
Boiling Pt: 260 °C
Melting Pt: 32…33 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005414
CAS Number: 492-97-7
EINECS: 207-767-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR