2-Amino-5-chloropyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Chloropyridin-2-amine, 5-Chloro-2-pyridylamine

OR13201-100G
APOSOR13201-100GEA 23.2 EUR
APOSOR13201-100G
2-Amino-5-chloropyridine 98%
2-Amino-5-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 127…128 °C/11mmHg
Melting Pt: 135…138 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00006324
CAS Number: 1072-98-6
EINECS: 214-020-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR