8-Methoxyquinoline-5-sulphonyl chloride

Supplier: Apollo Scientific
Danger

OR13482-1G OR13482-250MG
APOSOR13482-1GEA 642 EUR
APOSOR13482-1G APOSOR13482-250MG
8-Methoxyquinoline-5-sulphonyl chloride
8-Methoxyquinoline-5-sulphonyl chloride
Formula: C₁₀H₈ClNO₃S
MW: 257.7 g/mol
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD02752892
CAS Number: 90429-62-2
UN: 1759
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR