1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole

OR14885-250MG
APOSOR14885-250MGEA 39.3 EUR
APOSOR14885-250MG
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole 97%
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole
Formula: C₁₁H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD09817558
CAS Number: 62546-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR