Methyl-4-amino-3-nitrobenzoate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 4-amino-3-nitrobenzoate

OR15213-100G OR15213-25G OR15213-5G
APOSOR15213-100GEA 195 EUR
APOSOR15213-100G APOSOR15213-25G APOSOR15213-5G
Methyl-4-amino-3-nitrobenzoate
Methyl 4-amino-3-nitrobenzoate
Formula: C₈H₈N₂O₄
MW: 196.16 g/mol
Melting Pt: 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00017562
CAS Number: 3987-92-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR