N,N-Dimethyl(6-methylimidazo[1,2-a]pyridin-3-yl)methanamine

Supplier: Apollo Scientific
Warning

Synonyms: 3-[(Dimethylamino)methyl]-6-methylimidazo[1,2-a]pyridine

OR15401-1G OR15401-5G
APOSOR15401-1GEA 336 EUR
APOSOR15401-1G APOSOR15401-5G
N,N-Dimethyl(6-methylimidazo[1,2-a]pyridin-3-yl)methanamine
N,N-Dimethyl(6-methylimidazo[1,2-a]pyridin-3-yl)methanamine
Formula: C₁₁H₁₅N₃
MW: 189.26 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD06200990
CAS Number: 885949-46-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR