4-Bromo-2-ethoxythiazole

Supplier: Apollo Scientific
Warning

OR15556-1G
APOSOR15556-1GEA 40.6 EUR
APOSOR15556-1G
4-Bromo-2-ethoxythiazole
4-Bromo-2-ethoxythiazole
Formula: C₅H₆BrNOS
MW: 208.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787375
CAS Number: 240816-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR