2-Aminopyridine-4-boronic acid pinacol ester

Supplier: Apollo Scientific
Warning

Synonyms: 2-Aminopyridine-4-boronic acid, pinacol ester, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR17204-1G OR17204-100MG OR17204-5G
APOSOR17204-1GEA 241 EUR
APOSOR17204-1G APOSOR17204-100MG APOSOR17204-5G
2-Aminopyridine-4-boronic acid pinacol ester
2-Aminopyridine-4-boronic acid pinacol ester
Formula: C₁₁H₁₇BN₂O₂
MW: 220.08 g/mol
Melting Pt: 161…162 °C
Storage Temperature: Freezer
MDL Number: MFCD09607735
CAS Number: 1195995-72-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR