6-Chloro-N-hydroxynicotinamide

Supplier: Apollo Scientific

OR200102-1G OR200102-250MG OR200102-500MG OR200102-2.5G
APOSOR200102-1GEA 458 EUR
APOSOR200102-1G APOSOR200102-250MG APOSOR200102-500MG APOSOR200102-2.5G
6-Chloro-N-hydroxynicotinamide
6-Chloro-N-hydroxynicotinamide
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15144734
CAS Number: 1263094-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR