1,2,5-Trimethylpyrrole-3-carboxylic acid

Supplier: Apollo Scientific
Warning

OR23110-250MG
APOSOR23110-250MGEA 69.6 EUR
APOSOR23110-250MG
1,2,5-Trimethylpyrrole-3-carboxylic acid
1,2,5-Trimethylpyrrole-3-carboxylic acid
Formula: C₈H₁₁NO₂
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00203866
CAS Number: 175276-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR