Ethyl-4-(chlorosulphonyl)-2,5-dimethyl-3-furoate

Supplier: Apollo Scientific
Danger

OR2413-1G OR2413-250MG
APOSOR2413-1GEA 386 EUR
APOSOR2413-1G APOSOR2413-250MG
Ethyl-4-(chlorosulphonyl)-2,5-dimethyl-3-furoate
Ethyl-4-(chlorosulphonyl)-2,5-dimethyl-3-furoate
Formula: C₉H₁₁ClO₅S
MW: 266.7 g/mol
Boiling Pt: 110…114 °C/0.05mmHg
Storage Temperature: Ambient
MDL Number: MFCD01934807
CAS Number: 306936-32-3
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR