1-Adamantanecarbonitrile

Supplier: Apollo Scientific
Warning

Synonyms: 1-Cyanotricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decane-1-carbonitrile, 1-Cyanoadamantane

OR27249-250MG
APOSOR27249-250MGEA 58.2 EUR
APOSOR27249-250MG
1-Adamantanecarbonitrile
1-Adamantanecarbonitrile
Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Boiling Pt: 275
Melting Pt: 193…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00074731
CAS Number: 23074-42-2
UN: 3439
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR