Xanthene-9-carboxylic acid

Supplier: Apollo Scientific
Warning

Synonyms: 9H-Xanthene-9-carboxylic acid

OR27439-5G
APOSOR27439-5GEA 21.9 EUR
APOSOR27439-5G
Xanthene-9-carboxylic acid
Xanthene-9-carboxylic acid
Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Melting Pt: 221…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005059
CAS Number: 82-07-5
EINECS: 201-394-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR