2-Aminobenzenesulphonamide 98%

Supplier: Apollo Scientific
Warning

OR27532-25G OR27532-5G OR27532-1G
APOSOR27532-25GEA 105 EUR
APOSOR27532-25G APOSOR27532-5G APOSOR27532-1G
2-Aminobenzenesulphonamide 98%
2-Aminobenzenesulphonamide
Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
Melting Pt: 153…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00007932
CAS Number: 3306-62-5
EINECS: 221-988-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR