3-Benzyl-5-[(benzyloxy)methyl]-2-phenyl-1,3-oxazolidine

Supplier: Apollo Scientific

OR28271-25MG
APOSOR28271-25MGEA 21.9 EUR
APOSOR28271-25MG
3-Benzyl-5-[(benzyloxy)methyl]-2-phenyl-1,3-oxazolidine
3-Benzyl-5-[(benzyloxy)methyl]-2-phenyl-1,3-oxazolidine
Formula: C₂₄H₂₅NO₂
MW: 359.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01566985
CAS Number: 680214-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR