3-Chloro-4-hydroxybenzoic acid hemihydrate

Supplier: Apollo Scientific
Warning

OR2925-100G OR2925-500G OR2925-50G
APOSOR2925-100GEA 271 EUR
APOSOR2925-100G APOSOR2925-500G APOSOR2925-50G
3-Chloro-4-hydroxybenzoic acid hemihydrate
3-Chloro-4-hydroxybenzoic acid hemihydrate
Formula: 2C₇H₅ClO₃·H₂O
MW: 363.147 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD02662353
CAS Number: 128161-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR