1-(Quinolin-8-yl)ethanone

Supplier: Apollo Scientific
Warning

OR300272-1G OR300272-250MG OR300272-5G
APOSOR300272-1GEA 284 EUR
APOSOR300272-1G APOSOR300272-250MG APOSOR300272-5G
1-(Quinolin-8-yl)ethanone
1-(Quinolin-8-yl)ethanone
Formula: C₁₁H₉NO
MW: 171.195 g/mol
Boiling Pt: 176 °C (12 mm Hg)
Melting Pt: 42…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00047620
CAS Number: 56234-20-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR