2-Aminobenzothiazole-6-carbonitrile

Supplier: Apollo Scientific
Warning

OR300670-1G OR300670-5G
APOSOR300670-1GEA 85.7 EUR
APOSOR300670-1G APOSOR300670-5G
2-Aminobenzothiazole-6-carbonitrile
2-Aminobenzothiazole-6-carbonitrile
Formula: C₈H₅N₃S
MW: 175.2104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664359
CAS Number: 19759-66-1
UN: 3439
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR