2-Methyl-2-(methylamino)propan-1-ol

Supplier: Apollo Scientific
Warning

Synonyms: N,2-Dimethyl-1-hydroxypropan-2-amine, 1-Hydroxy-2-methyl-2-(methylamino)propane

OR301186-1G OR301186-5G
APOSOR301186-1GEA 109 EUR
APOSOR301186-1G APOSOR301186-5G
2-Methyl-2-(methylamino)propan-1-ol
2-Methyl-2-(methylamino)propan-1-ol
Formula: C₅H₁₃NO
MW: 103.163 g/mol
Boiling Pt: 162 °C
Density: 0.878 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD12184529
CAS Number: 27646-80-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR