5-Methyl-1,3,4-oxadiazol-2-amine 98%

Supplier: Apollo Scientific

OR301230-5G
APOSOR301230-5GEA 63.8 EUR
APOSOR301230-5G
5-Methyl-1,3,4-oxadiazol-2-amine 98%
5-Methyl-1,3,4-oxadiazol-2-amine
Formula: C₃H₅N₃O
MW: 99.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03703470
CAS Number: 52838-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR