2-Amino-5-bromo-3-pyridinol 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-5-bromo-3-hydroxypyridine

OR301243-25G OR301243-5G OR301243-100G
APOSOR301243-25GEA 220 EUR
APOSOR301243-25G APOSOR301243-5G APOSOR301243-100G
2-Amino-5-bromo-3-pyridinol 98%
2-Amino-5-bromo-3-pyridinol
Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09744143
CAS Number: 39903-01-0
EINECS: 689-734-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR