7-(Hydroxymethyl)-2-azaspiro[4.4]nonan-3-one

Supplier: Apollo Scientific

OR304439-1G
APOSOR304439-1GEA 3380 EUR
APOSOR304439-1G
7-(Hydroxymethyl)-2-azaspiro[4.4]nonan-3-one
7-(Hydroxymethyl)-2-azaspiro[4.4]nonan-3-one
Formula: C₉H₁₅NO₂
MW: 169.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28649465
CAS Number: 1782480-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR