5-Bromo-6-nitro-1H-indazole 97%

Supplier: Apollo Scientific

OR305536-1G
APOSOR305536-1GEA 66.7 EUR
APOSOR305536-1G
5-Bromo-6-nitro-1H-indazole 97%
5-Bromo-6-nitro-1H-indazole
Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027584
CAS Number: 71785-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR