4-Bromo-2-methylisoindolin-1-one

Supplier: Apollo Scientific
Warning

Synonyms: 4-Bromo-2,3-dihydro-2-methyl-1H-isoindol-1-one, 4-Bromo-2,3-dihydro-2-methyl-1-oxo-1H-isoindole

OR30752-250MG
APOSOR30752-250MGEA 102 EUR
APOSOR30752-250MG
4-Bromo-2-methylisoindolin-1-one
4-Bromo-2-methylisoindolin-1-one
Formula: C₉H₈BrNO
MW: 226.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11878013
CAS Number: 435273-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR