1-Amino-2-(4-methoxy-phenyl)-propan-2-ol hydrochloride

Supplier: Apollo Scientific

OR307628-1G
APOSOR307628-1GEA 451 EUR
APOSOR307628-1G
1-Amino-2-(4-methoxy-phenyl)-propan-2-ol hydrochloride
1-Amino-2-(4-methoxy-phenyl)-propan-2-ol hydrochloride
Formula: C₁₀H₁₅NO₂·ClH
MW: 217.693 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366615
CAS Number: 1172878-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR