4-Chloro-6,8-dibromo-2-methylquinoline

Supplier: Apollo Scientific

OR307820-1G
APOSOR307820-1GEA 375 EUR
APOSOR307820-1G
4-Chloro-6,8-dibromo-2-methylquinoline
4-Chloro-6,8-dibromo-2-methylquinoline
Formula: C₁₀H₆Br₂ClN
MW: 335.422 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272436
CAS Number: 203626-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR