4-Chloro-6-ethoxyquinoline

Supplier: Apollo Scientific

OR307967-1G OR307967-5G
APOSOR307967-1GEA 325 EUR
APOSOR307967-1G APOSOR307967-5G
4-Chloro-6-ethoxyquinoline
4-Chloro-6-ethoxyquinoline
Formula: C₁₁H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD09787733
CAS Number: 103862-63-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR