4-Amino-2-propylquinoline

Supplier: Apollo Scientific

OR309309-1G
APOSOR309309-1GEA 375 EUR
APOSOR309309-1G
4-Amino-2-propylquinoline
4-Amino-2-propylquinoline
Formula: C₁₂H₁₄N₂
MW: 186.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12674938
CAS Number: 57165-80-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR