6-Bromo-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride

Supplier: Apollo Scientific

OR309377-1G
APOSOR309377-1GEA 335 EUR
APOSOR309377-1G
6-Bromo-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride
6-Bromo-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride
Formula: C₁₄H₁₆BrN₃·ClH
MW: 342.662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20036778
CAS Number: 1333256-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR