2-Amino-5-iodo-4,6-dimethylpyridine 95%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Iodo-4,6-dimethylpyridin-2-amine

OR310150-1G
APOSOR310150-1GEA 206 EUR
APOSOR310150-1G
2-Amino-5-iodo-4,6-dimethylpyridine 95%
2-Amino-5-iodo-4,6-dimethylpyridine
Formula: C₇H₉IN₂
MW: 248.07 g/mol
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD06798226
CAS Number: 885952-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR