6-Bromo-2-oxo-1,2-dihydroquinoline-4-carboxylic acid

Supplier: Apollo Scientific
Warning

OR310688-1G
APOSOR310688-1GEA 169 EUR
APOSOR310688-1G
6-Bromo-2-oxo-1,2-dihydroquinoline-4-carboxylic acid
6-Bromo-2-oxo-1,2-dihydroquinoline-4-carboxylic acid
Formula: C₁₀H₆BrNO₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD09837171
CAS Number: 5463-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR