Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate 97%

Supplier: Apollo Scientific
Warning

OR312473-1G OR312473-5G
APOSOR312473-1GEA 154 EUR
APOSOR312473-1G APOSOR312473-5G
Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate 97%
Methyl 3-((tert-butoxycarbonyl)amino)bicyclo[1.1.1]pentane-1-carboxylate
Formula: C₁₂H₁₉NO₄
MW: 241.284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24476148
CAS Number: 676371-64-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR