1-Boc-4-(aminomethyl)-4-hydroxypiperidine 95%

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate, 1-Boc-4-(Aminomethyl)-4-hydroxypiperidine

OR315339-5G
APOSOR315339-5GEA 144 EUR
APOSOR315339-5G
1-Boc-4-(aminomethyl)-4-hydroxypiperidine 95%
1-Boc-4-(aminomethyl)-4-hydroxypiperidine
Formula: C₁₁H₂₂N₂O₃
MW: 230.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11579803
CAS Number: 392331-66-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR