cis-3-Hydroxycyclobutanecarboxylic acid

Supplier: Apollo Scientific

OR317124-1G
APOSOR317124-1GEA 88.6 EUR
APOSOR317124-1G
cis-3-Hydroxycyclobutanecarboxylic acid
cis-3-Hydroxycyclobutanecarboxylic acid
Formula: C₅H₈O₃
MW: 116.1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12964956
CAS Number: 552849-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR