N'-[(1E)-1-(4-Bromophenyl)ethylidene]methoxycarbohydrazide, tech

Supplier: Apollo Scientific

OR32068-1G
APOSOR32068-1GEA 1570 EUR
APOSOR32068-1G
N'-[(1E)-1-(4-Bromophenyl)ethylidene]methoxycarbohydrazide, tech
N'-[(1E)-1-(4-Bromophenyl)ethylidene]methoxycarbohydrazide
Formula: C₁₀H₁₁BrN₂O₂
MW: 271.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00792940
CAS Number: 1211461-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR