tert-Butyl (S)-2-ethylpiperidine-1-carboxylate 97%

Supplier: Apollo Scientific
Warning

OR321206-100MG
APOSOR321206-100MGEA 168 EUR
APOSOR321206-100MG
tert-Butyl (S)-2-ethylpiperidine-1-carboxylate 97%
tert-Butyl (S)-2-ethylpiperidine-1-carboxylate
Formula: C₁₂H₂₃NO₂
MW: 213.317 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28355903
CAS Number: 1853230-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR