4-Hydroxy-3-(1,2,3,4-tetrahydroisoquinolin-1-yl)-2H-chromen-2-one, tech

Supplier: Apollo Scientific

OR32146-1G
APOSOR32146-1GEA 1570 EUR
APOSOR32146-1G
4-Hydroxy-3-(1,2,3,4-tetrahydroisoquinolin-1-yl)-2H-chromen-2-one, tech
4-Hydroxy-3-(1,2,3,4-tetrahydroisoquinolin-1-yl)-2H-chromen-2-one
Formula: C₁₈H₁₅NO₃
MW: 293.317 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05974927
CAS Number: 860649-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR