5-Chloro-6-methyl-2-phenylpyrimidin-4-ol 95%

Supplier: Apollo Scientific

OR32952-1G
APOSOR32952-1GEA 1120 EUR
APOSOR32952-1G
5-Chloro-6-methyl-2-phenylpyrimidin-4-ol 95%
5-Chloro-6-methyl-2-phenylpyrimidin-4-ol
Formula: C₁₁H₉ClN₂O
MW: 220.655 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00232127
CAS Number: 20551-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR