5-Cyclopropanecarbonyl-2-cyclopropylpyridine, tech

Supplier: Apollo Scientific

OR33046-1G
APOSOR33046-1GEA 1570 EUR
APOSOR33046-1G
5-Cyclopropanecarbonyl-2-cyclopropylpyridine, tech
5-Cyclopropanecarbonyl-2-cyclopropylpyridine
Formula: C₁₂H₁₃NO
MW: 187.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05256200
CAS Number: 56260-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR