1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-3,3-dimethylazetidin-2-one, tech

Supplier: Apollo Scientific

OR33242-1G
APOSOR33242-1GEA 1570 EUR
APOSOR33242-1G
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-3,3-dimethylazetidin-2-one, tech
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-3,3-dimethylazetidin-2-one
Formula: C₁₆H₂₀N₂O₂
MW: 272.342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00793419
CAS Number: 339100-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR