4-Bromo-2-methyl-3-quinolinol 95%

Supplier: Apollo Scientific

OR33495-1G
APOSOR33495-1GEA 209 EUR
APOSOR33495-1G
4-Bromo-2-methyl-3-quinolinol 95%
4-Bromo-2-methyl-3-quinolinol
Formula: C₁₀H₈BrNO
MW: 238.08
Storage Temperature: Ambient
CAS Number: 13235-12-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR