2-Acetyl-4-methylpyridine 98%

Supplier: Apollo Scientific

Synonyms: 1-(4-Methylpyridin-2-yl)ethanone

OR345116-5G
APOSOR345116-5GEA 68.3 EUR
APOSOR345116-5G
2-Acetyl-4-methylpyridine 98%
2-Acetyl-4-methylpyridine
Formula: C₈H₉NO
MW: 135.1632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863638
CAS Number: 59576-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR