2-Nitropyridine-3-carboxylic acid 97%

Supplier: Apollo Scientific

Synonyms: 2-Nitronicotinic acid

OR345320-1G
APOSOR345320-1GEA 95.7 EUR
APOSOR345320-1G
2-Nitropyridine-3-carboxylic acid 97%
2-Nitropyridine-3-carboxylic acid
Formula: C₆H₄N₂O₄
MW: 168.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114130
CAS Number: 33225-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR