2-Amino-5-chloropyridine-3-sulphonyl chloride

Supplier: Apollo Scientific
Danger

OR346130-1G OR346130-250MG OR346130-5G
APOSOR346130-1GEA 227 EUR
APOSOR346130-1G APOSOR346130-250MG APOSOR346130-5G
2-Amino-5-chloropyridine-3-sulphonyl chloride
2-Amino-5-chloropyridine-3-sulphonyl chloride
Formula: C₅H₄Cl₂N₂O₂S
MW: 227.07 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD12402514
CAS Number: 163137-45-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR