2-(3,4-Dimethoxyphenyl)-3-methylbutanenitrile

Supplier: Apollo Scientific
Warning

Synonyms: Verapamil Impurity K

OR346216-1G OR346216-5G
APOSOR346216-1GEA 0 EUR
APOSOR346216-1G APOSOR346216-5G
2-(3,4-Dimethoxyphenyl)-3-methylbutanenitrile
2-(3,4-Dimethoxyphenyl)-3-methylbutanenitrile
Formula: C₁₃H₁₇NO₂
MW: 219.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800230
CAS Number: 20850-49-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR