3,4-Dimethyldiphenylamine

Supplier: Apollo Scientific

Synonyms: Phenyl(3,4-xylyl)amine, N-Phenyl-3,4-xylidine

OR350141-25G
APOSOR350141-25GEA 26.1 EUR
APOSOR350141-25G
3,4-Dimethyldiphenylamine
3,4-Dimethyldiphenylamine
Formula: C14H15N
MW: 197.2800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093248
CAS Number: 17802-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR