9-Bromo-7-methyl-2-morpholino-4H-pyrido[1,2-a]pyrimidin-4-one ≥98%

Supplier: Apollo Scientific

OR350434-100MG
APOSOR350434-100MGEA 201 EUR
APOSOR350434-100MG
9-Bromo-7-methyl-2-morpholino-4H-pyrido[1,2-a]pyrimidin-4-one ≥98%
9-Bromo-7-methyl-2-morpholino-4H-pyrido[1,2-a]pyrimidin-4-one
Formula: C₁₃H₁₄BrN₃O₂
MW: 324.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113210
CAS Number: 351002-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR