2,2-Dimethylethenylboronic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: (2-Methylprop-1-en-1-yl)boronic acid

OR360167-1G
APOSOR360167-1GEA 26.1 EUR
APOSOR360167-1G
2,2-Dimethylethenylboronic acid 95%
2,2-Dimethylethenylboronic acid
Formula: C₄H₉BO₂
MW: 99.93 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01074609
CAS Number: 14559-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR