4-(N,N-Dimethylamino)phenylboronic acid pinacol ester

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Dimethylamino)phenylboronic acid pinacol ester, N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

OR360226-25G
APOSOR360226-25GEA 79.9 EUR
APOSOR360226-25G
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester
Formula: C₁₄H₂₂BNO₂
MW: 247.1410 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD05663854
CAS Number: 171364-78-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR