5-Chloro-2-tetralone ≥98%

Supplier: Apollo Scientific

OR370117-100MG
APOSOR370117-100MGEA 173 EUR
APOSOR370117-100MG
5-Chloro-2-tetralone ≥98%
5-Chloro-2-tetralone
Formula: C₁₀H₉ClO
MW: 180.631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038574
CAS Number: 69739-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR